2,2-Difluoroethyl 3-fluoropropyl carbonate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC450302-25G
APOSPC450302-25GEA 1550 EUR
APOSPC450302-25G
2,2-Difluoroethyl 3-fluoropropyl carbonate
2,2-Difluoroethyl 3-fluoropropyl carbonate
Formule: C₆H₉F₃O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28334296
CAS nummer: 1980044-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR