2,2,3,3-Tetrafluoropropyl 1H,1H-perfluorohexyl carbonate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC450340-25G
APOSPC450340-25GEA 1550 EUR
APOSPC450340-25G
2,2,3,3-Tetrafluoropropyl 1H,1H-perfluorohexyl carbonate
2,2,3,3-Tetrafluoropropyl 1H,1H-perfluorohexyl carbonate
Formule: C₁₀H₅F₁₅O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28334333
CAS nummer: 1980040-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR