Bis(1H,1H,7H-perfluoroheptyl) carbonate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC450377-25G
APOSPC450377-25GEA 1550 EUR
APOSPC450377-25G
Bis(1H,1H,7H-perfluoroheptyl) carbonate
Bis(1H,1H,7H-perfluoroheptyl) carbonate
Formule: C₁₅H₆F₂₄O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00153659
CAS nummer: 866-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR