tert-Butyl 2,2,3,3,3-pentafluoropropyl carbonate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC450459-25G
APOSPC450459-25GEA 1550 EUR
APOSPC450459-25G
tert-Butyl 2,2,3,3,3-pentafluoropropyl carbonate
tert-Butyl 2,2,3,3,3-pentafluoropropyl carbonate
Formule: C₈H₁₁F₅O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28334432
CAS nummer: 1980035-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR