N-Cyclohexyl-2,2,5,5-tetrakis(trifluoromethyl)-1,3-dioxolan-4-imine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC450488-1G PC450488-250MG
APOSPC450488-1GEA 926 EUR
APOSPC450488-1G APOSPC450488-250MG
N-Cyclohexyl-2,2,5,5-tetrakis(trifluoromethyl)-1,3-dioxolan-4-imine 95%
N-Cyclohexyl-2,2,5,5-tetrakis(trifluoromethyl)-1,3-dioxolan-4-imine
Formule: C₁₃H₁₁F₁₂NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD29765768
CAS nummer: 5771-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR