2,2,3,4,4,4-Hexafluorobutyl methacrylate 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC4657-100G PC4657-25G PC4657-5G
APOSPC4657-100GEA 242 EUR
APOSPC4657-100G APOSPC4657-25G APOSPC4657-5G
2,2,3,4,4,4-Hexafluorobutyl methacrylate 98%
2,2,3,4,4,4-Hexafluorobutyl methacrylate

Stabilised; Refrigeration will extend shelf-life.

Formule: C₈H₈F₆O₂
Kookpunt: 158 °C
Dichtheid: 1,348 g/cm³ (25 °C)
Vlampunt: 57 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00042311
CAS nummer: 36405-47-7
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR