N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine 90%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC4728-100G PC4728-25G PC4728-5G
APOSPC4728-100GEA 820 EUR
APOSPC4728-100G APOSPC4728-25G APOSPC4728-5G
N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine 90%
N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine
Formule: C₆H₁₁ClF₃N
Kookpunt: 38 °C/10mmHg
Dichtheid: 1,19 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00054672
CAS nummer: 357-83-5
UN: 3267
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR