4'-Chloro-5-fluoro-1,1'-biphenyl-2-amine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC48313-1G PC48313-100MG PC48313-250MG PC48313-500MG PC48313-2.5G PC48313-5G
APOSPC48313-1GEA 368 EUR
APOSPC48313-1G APOSPC48313-100MG APOSPC48313-250MG APOSPC48313-500MG APOSPC48313-2.5G APOSPC48313-5G
4'-Chloro-5-fluoro-1,1'-biphenyl-2-amine
4'-Chloro-5-fluoro-1,1'-biphenyl-2-amine
Formule: C₁₂H₉ClFN
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD12824033
CAS nummer: 188731-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR