2-(Difluoromethyl)benzoic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC48345-1G PC48345-250MG PC48345-5G
APOSPC48345-1GEA 43.8 EUR
APOSPC48345-1G APOSPC48345-250MG APOSPC48345-5G
2-(Difluoromethyl)benzoic acid 97%
2-(Difluoromethyl)benzoic acid
Formule: C₈H₆F₂O₂
Smeltpunt: 128…130 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16140191
CAS nummer: 799814-32-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR