1-(3-Bromo-4-fluorophenyl)-3-(2-chloroethyl)urea

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC48674-1G PC48674-250MG
APOSPC48674-1GEA 210 EUR
APOSPC48674-1G APOSPC48674-250MG
1-(3-Bromo-4-fluorophenyl)-3-(2-chloroethyl)urea
1-(3-Bromo-4-fluorophenyl)-3-(2-chloroethyl)urea
Formule: C₉H₉BrClFN₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD31372792
CAS nummer: 2168111-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR