2-Hydroxy-3-(trifluoromethyl)pyridine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC4868W-100G PC4868W-10G PC4868W-25G
APOSPC4868W-100GEA 77.5 EUR
APOSPC4868W-100G APOSPC4868W-10G APOSPC4868W-25G
2-Hydroxy-3-(trifluoromethyl)pyridine 97%
2-Hydroxy-3-(trifluoromethyl)pyridine
Formule: C₆H₄F₃NO
Smeltpunt: 150…155 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00153194
CAS nummer: 22245-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR