6-Bromo-8-fluoroquinoline 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC49040-1G PC49040-5G PC49040-25G
APOSPC49040-1GEA 43.8 EUR
APOSPC49040-1G APOSPC49040-5G APOSPC49040-25G
6-Bromo-8-fluoroquinoline 95%
6-Bromo-8-fluoroquinoline
Formule: C₉H₅BrFN
Smeltpunt: 103…106 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11053791
CAS nummer: 220513-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR