5-Ethynyl-2-(trifluoromethyl)pyridine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC49132-1G PC49132-250MG PC49132-5G
APOSPC49132-1GEA 168 EUR
APOSPC49132-1G APOSPC49132-250MG APOSPC49132-5G
5-Ethynyl-2-(trifluoromethyl)pyridine 95%
5-Ethynyl-2-(trifluoromethyl)pyridine
Formule: C₈H₄F₃N
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD13189725
CAS nummer: 1196155-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR