2-Amino-5-(trifluoromethoxy)benzonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC49304-1G PC49304-5G
APOSPC49304-1GEA 189 EUR
APOSPC49304-1G APOSPC49304-5G
2-Amino-5-(trifluoromethoxy)benzonitrile
2-Amino-5-(trifluoromethoxy)benzonitrile
Formule: C₈H₅F₃N₂O
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD06408813
CAS nummer: 549488-77-9
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR