4-Bromo-2-fluorophenylacetylene

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC49319-1G PC49319-250MG
APOSPC49319-1GEA 58.4 EUR
APOSPC49319-1G APOSPC49319-250MG
4-Bromo-2-fluorophenylacetylene
4-Bromo-2-fluorophenylacetylene
Formule: C₈H₄BrF
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD02683523
CAS nummer: 302912-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR