2-Methyl-4-(trifluoromethoxy)benzylamine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: [2-Methyl-4-(trifluoromethoxy)phenyl]methylamine, 2-(Aminomethyl)-5-(trifluoromethoxy)toluene

PC49352-1G PC49352-250MG
APOSPC49352-1GEA 176 EUR
APOSPC49352-1G APOSPC49352-250MG
2-Methyl-4-(trifluoromethoxy)benzylamine
2-Methyl-4-(trifluoromethoxy)benzylamine
Formule: C₉H₁₀F₃NO
MW: 205.177 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06213547
CAS nummer: 771572-39-5
UN: 3259
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR