2-Bromo-4,5-difluorobenzyl alcohol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC49498-1G PC49498-5G
APOSPC49498-1GEA 123 EUR
APOSPC49498-1G APOSPC49498-5G
2-Bromo-4,5-difluorobenzyl alcohol
2-Bromo-4,5-difluorobenzyl alcohol
Formule: C₇H₅BrF₂O
Smeltpunt: 75…76 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18909667
CAS nummer: 476620-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR