3-(Trifluoromethyl)piperidine 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC49500-25G PC49500-5G
APOSPC49500-25GEA 615 EUR
APOSPC49500-25G APOSPC49500-5G
3-(Trifluoromethyl)piperidine 98%
3-(Trifluoromethyl)piperidine
Formule: C₆H₁₀F₃N
Kookpunt: 74…75 °C/33mmHg
Dichtheid: 1,154 g/cm³ (25 °C)
Vlampunt: 28 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD02183561
CAS nummer: 768-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR