2,4-Difluoro-5-hydroxybenzotrifluoride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC500224-1G PC500224-250MG
APOSPC500224-1GEA 555 EUR
APOSPC500224-1G APOSPC500224-250MG
2,4-Difluoro-5-hydroxybenzotrifluoride
2,4-Difluoro-5-hydroxybenzotrifluoride
Formule: C₇H₃F₅O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD27937185
CAS nummer: 1632283-20-5
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR