4'-Bromo-5'-chloro-2'-fluoroacetophenone

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC500405-1G PC500405-250MG
APOSPC500405-1GEA 438 EUR
APOSPC500405-1G APOSPC500405-250MG
4'-Bromo-5'-chloro-2'-fluoroacetophenone
4'-Bromo-5'-chloro-2'-fluoroacetophenone
Formule: C₈H₅BrClFO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD27995573
CAS nummer: 1594605-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR