3-Bromo-6-chloro-2-fluorobenzoic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC500462-1G PC500462-25G PC500462-5G
APOSPC500462-1GEA 46.8 EUR
APOSPC500462-1G APOSPC500462-25G APOSPC500462-5G
3-Bromo-6-chloro-2-fluorobenzoic acid
3-Bromo-6-chloro-2-fluorobenzoic acid
Formule: C₇H₃BrClFO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11110256
CAS nummer: 702640-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR