5-(Bromomethyl)-2-fluorobenzoic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC500479-1G PC500479-250MG PC500479-5G
APOSPC500479-1GEA 330 EUR
APOSPC500479-1G APOSPC500479-250MG APOSPC500479-5G
5-(Bromomethyl)-2-fluorobenzoic acid
5-(Bromomethyl)-2-fluorobenzoic acid
Formule: C₈H₆BrFO₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD06208849
CAS nummer: 773100-76-8
UN: 3261
ADR: 8,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR