2-(Bromomethyl)-3-fluorobenzonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC500524-1G PC500524-250MG PC500524-5G
APOSPC500524-1GEA 67.2 EUR
APOSPC500524-1G APOSPC500524-250MG APOSPC500524-5G
2-(Bromomethyl)-3-fluorobenzonitrile
2-(Bromomethyl)-3-fluorobenzonitrile
Formule: C₈H₅BrFN
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD12025416
CAS nummer: 635723-84-1
UN: 3261
ADR: 8,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR