4-Fluoro-3-(trifluoromethoxy)aniline 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC500550-1G PC500550-5G PC500550-25G
APOSPC500550-1GEA 78.9 EUR
APOSPC500550-1G APOSPC500550-5G APOSPC500550-25G
4-Fluoro-3-(trifluoromethoxy)aniline 98%
4-Fluoro-3-(trifluoromethoxy)aniline
Formule: C₇H₅F₄NO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09832246
CAS nummer: 113421-98-0
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR