Methyl 4-cyano-2-fluorobenzoate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC500686-25G PC500686-5G PC500686-50G
APOSPC500686-25GEA 347 EUR
APOSPC500686-25G APOSPC500686-5G APOSPC500686-50G
Methyl 4-cyano-2-fluorobenzoate
Methyl 4-cyano-2-fluorobenzoate
Formule: C₉H₆FNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04335187
CAS nummer: 175596-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR