3-Fluoro-2-nitrobenzyl alcohol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC501154-1G PC501154-250MG PC501154-5G
APOSPC501154-1GEA 46.8 EUR
APOSPC501154-1G APOSPC501154-250MG APOSPC501154-5G
3-Fluoro-2-nitrobenzyl alcohol 95%
3-Fluoro-2-nitrobenzyl alcohol
Formule: C₇H₆FNO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD13185620
CAS nummer: 1214323-11-1

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR