3'-Fluoro-4'-(trifluoromethoxy)acetophenone 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC501196-1G PC501196-5G PC501196-25G
APOSPC501196-1GEA 116 EUR
APOSPC501196-1G APOSPC501196-5G APOSPC501196-25G
3'-Fluoro-4'-(trifluoromethoxy)acetophenone 99%
3'-Fluoro-4'-(trifluoromethoxy)acetophenone
Formule: C₉H₆F₄O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD15142928
CAS nummer: 1174006-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR