2-Bromo-3-fluoro-4-iodoaniline

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC501459-1G PC501459-250MG PC501459-5G
APOSPC501459-1GEA 476 EUR
APOSPC501459-1G APOSPC501459-250MG APOSPC501459-5G
2-Bromo-3-fluoro-4-iodoaniline
2-Bromo-3-fluoro-4-iodoaniline
Formule: C₆H₄BrFIN
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28397846
CAS nummer: 1936364-85-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR