2-Bromo-4,6-difluorobenzoic acid 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC501567-10G PC501567-1G PC501567-5G
APOSPC501567-10GEA 1170 EUR
APOSPC501567-10G APOSPC501567-1G APOSPC501567-5G
2-Bromo-4,6-difluorobenzoic acid 95%
2-Bromo-4,6-difluorobenzoic acid
Formule: C₇H₃BrF₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD23713104
CAS nummer: 651026-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR