6-Bromo-2,3-difluorobenzyl bromide 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC501975-10G PC501975-1G PC501975-5G
APOSPC501975-10GEA 1250 EUR
APOSPC501975-10G APOSPC501975-1G APOSPC501975-5G
6-Bromo-2,3-difluorobenzyl bromide 97%
6-Bromo-2,3-difluorobenzyl bromide
Formule: C₇H₄Br₂F₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28060860
CAS nummer: 1807193-48-1

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR