2,4-Difluoro-3-iodobenzotrifluoride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC502070-1G PC502070-250MG PC502070-5G
APOSPC502070-1GEA 483 EUR
APOSPC502070-1G APOSPC502070-250MG APOSPC502070-5G
2,4-Difluoro-3-iodobenzotrifluoride
2,4-Difluoro-3-iodobenzotrifluoride
Formule: C₇H₂F₅I
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD30481111
CAS nummer: 2089257-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR