4-Fluoro-3-[(trifluoromethyl)sulphonyl]benzenesulphonyl chloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 4-Fluoro-3-((trifluoromethyl)sulphonyl)benzene-1-sulphonyl chloride

PC502180-1G PC502180-250MG PC502180-5G
APOSPC502180-1GEA 80.4 EUR
APOSPC502180-1G APOSPC502180-250MG APOSPC502180-5G
4-Fluoro-3-[(trifluoromethyl)sulphonyl]benzenesulphonyl chloride
4-Fluoro-3-[(trifluoromethyl)sulphonyl]benzenesulphonyl chloride
Formule: C₇H₃ClF₄O₄S₂
MW: 326.673 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16883063
CAS nummer: 1027345-07-8
UN: 1760
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR