3-Bromo-2-hydroxy-5-(trifluoromethoxy)benzaldehyde 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC502224-1G PC502224-250MG PC502224-5G
APOSPC502224-1GEA 58.4 EUR
APOSPC502224-1G APOSPC502224-250MG APOSPC502224-5G
3-Bromo-2-hydroxy-5-(trifluoromethoxy)benzaldehyde 95%
3-Bromo-2-hydroxy-5-(trifluoromethoxy)benzaldehyde
Formule: C₈H₄BrF₃O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD30730320
CAS nummer: 2149601-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR