2,3-Difluoro-6-(trifluoromethoxy)benzaldehyde 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC502497-1G
APOSPC502497-1GEA 577 EUR
APOSPC502497-1G
2,3-Difluoro-6-(trifluoromethoxy)benzaldehyde 95%
2,3-Difluoro-6-(trifluoromethoxy)benzaldehyde
Formule: C₈H₃F₅O₂
MDL Nummer: MFCD31561166
CAS nummer: 2244081-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR