2-Bromo-6-(trifluoromethoxy)benzaldehyde

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC502684-1G PC502684-5G PC502684-10G
APOSPC502684-1GEA 61.4 EUR
APOSPC502684-1G APOSPC502684-5G APOSPC502684-10G
2-Bromo-6-(trifluoromethoxy)benzaldehyde
2-Bromo-6-(trifluoromethoxy)benzaldehyde
Formule: C₈H₄BrF₃O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11856022
CAS nummer: 1114809-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR