2-Bromo-6-(trifluoromethoxy)benzyl alcohol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC502688-1G PC502688-5G PC502688-10G
APOSPC502688-1GEA 132 EUR
APOSPC502688-1G APOSPC502688-5G APOSPC502688-10G
2-Bromo-6-(trifluoromethoxy)benzyl alcohol 95%
2-Bromo-6-(trifluoromethoxy)benzyl alcohol
Formule: C₈H₆BrF₃O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD22375105
CAS nummer: 1253189-03-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR