4-Bromo-2,3-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC502797-1G
APOSPC502797-1GEA 1180 EUR
APOSPC502797-1G
4-Bromo-2,3-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
4-Bromo-2,3-difluorophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formule: C₁₃H₄BrF₇O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD31630015
CAS nummer: 2244084-87-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR