4-Fluoro-3-methyl-5-(trifluoromethyl)nitrobenzene 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC50421-10G PC50421-1G PC50421-5G
APOSPC50421-10GEA 593 EUR
APOSPC50421-10G APOSPC50421-1G APOSPC50421-5G
4-Fluoro-3-methyl-5-(trifluoromethyl)nitrobenzene 97%
4-Fluoro-3-methyl-5-(trifluoromethyl)nitrobenzene
Formule: C₈H₅F₄NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28797683
CAS nummer: 1803832-55-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR