Methyl 2-hydrazino-4-(trifluoromethyl)pyrimidine-5-carboxylate 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC5048-100MG
APOSPC5048-100MGEA 29.2 EUR
APOSPC5048-100MG
Methyl 2-hydrazino-4-(trifluoromethyl)pyrimidine-5-carboxylate 95%
Methyl 2-hydrazino-4-(trifluoromethyl)pyrimidine-5-carboxylate
Formule: C₇H₇F₃N₄O₂
Smeltpunt: 132…134 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00068147
CAS nummer: 175137-28-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR