1-[2-Bromo-4-(trifluoromethyl)phenyl]-3-(2-chloroethyl)urea

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC50539-1G PC50539-250MG
APOSPC50539-1GEA 26.3 EUR
APOSPC50539-1G APOSPC50539-250MG
1-[2-Bromo-4-(trifluoromethyl)phenyl]-3-(2-chloroethyl)urea
1-[2-Bromo-4-(trifluoromethyl)phenyl]-3-(2-chloroethyl)urea
Formule: C₁₀H₉BrClF₃N₂O
MW: 345.543 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD32264076
CAS nummer: 2407339-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR