2-Fluoro-4-(methylsulphonyl)phenylboronic acid pinacol ester

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC510141-1G
APOSPC510141-1GEA 645 EUR
APOSPC510141-1G
2-Fluoro-4-(methylsulphonyl)phenylboronic acid pinacol ester
2-Fluoro-4-(methylsulphonyl)phenylboronic acid pinacol ester
Formule: C₁₃H₁₈BFO₄S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD18760181
CAS nummer: 1384951-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR