Potassium Trifluoro[3-(methoxymethoxy)phenyl]borate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC510188-1G
APOSPC510188-1GEA 922 EUR
APOSPC510188-1G
Potassium Trifluoro[3-(methoxymethoxy)phenyl]borate
Potassium Trifluoro[3-(methoxymethoxy)phenyl]borate
Formule: C₈H₉BF₃O₂·K
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD21223197
CAS nummer: 1542163-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR