3-[(4-Fluorophenyl)(hydroxy)methyl]isoxazole-4-carboxylic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC510272-1G
APOSPC510272-1GEA 1230 EUR
APOSPC510272-1G
3-[(4-Fluorophenyl)(hydroxy)methyl]isoxazole-4-carboxylic acid
3-[(4-Fluorophenyl)(hydroxy)methyl]isoxazole-4-carboxylic acid
Formule: C₁₁H₈FNO₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28977389
CAS nummer: 2006277-43-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR