5-Chloro-6-fluoroisatin

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC510278-1G
APOSPC510278-1GEA 730 EUR
APOSPC510278-1G
5-Chloro-6-fluoroisatin
5-Chloro-6-fluoroisatin
Formule: C₈H₃ClFNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09728573
CAS nummer: 953897-06-8

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR