(1H,1H,2H,2H-Tridecafluorooct-1-yl)phosphonic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC51063-1G PC51063-5G
APOSPC51063-1GEA 123 EUR
APOSPC51063-1G APOSPC51063-5G
(1H,1H,2H,2H-Tridecafluorooct-1-yl)phosphonic acid
(1H,1H,2H,2H-Tridecafluorooct-1-yl)phosphonic acid
Formule: C₈H₆F₁₃O₃P
Smeltpunt: 168…173 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD18910708
CAS nummer: 252237-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR