2-Fluoro-5-(isopropylcarbamoyl)phenylboronic acid 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC5111-1G
APOSPC5111-1GEA 177 EUR
APOSPC5111-1G
2-Fluoro-5-(isopropylcarbamoyl)phenylboronic acid 98%
2-Fluoro-5-(isopropylcarbamoyl)phenylboronic acid
Formule: C₁₀H₁₃BFNO₃
Smeltpunt: 200…203 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD08235103
CAS nummer: 874289-49-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR