2-Chloro-3-fluoro-4-hydroxypyridine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC51119-100MG PC51119-1G PC51119-5G
APOSPC51119-100MGEA 65.8 EUR
APOSPC51119-100MG APOSPC51119-1G APOSPC51119-5G
2-Chloro-3-fluoro-4-hydroxypyridine
2-Chloro-3-fluoro-4-hydroxypyridine
Formule: C₅H₃ClFNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD13189885
CAS nummer: 1184172-46-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR