2,3-Dichloro-6-fluorobenzaldehyde 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC51143-25G PC51143-5G PC51143-1G
APOSPC51143-25GEA 388 EUR
APOSPC51143-25G APOSPC51143-5G APOSPC51143-1G
2,3-Dichloro-6-fluorobenzaldehyde 97%
2,3-Dichloro-6-fluorobenzaldehyde
Formule: C₇H₃Cl₂FO
Smeltpunt: 56…60 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04115870
CAS nummer: 95399-95-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR