Methyl nonafluorobutyl ketone 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC5118-5G PC5118-25G
APOSPC5118-5GEA 162 EUR
APOSPC5118-5G APOSPC5118-25G
Methyl nonafluorobutyl ketone 98%
Methyl nonafluorobutyl ketone
Formule: C₆H₃F₉O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00153209
CAS nummer: 678-18-2
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR