5-Bromo-3-fluoro-2-hydroxybenzoic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC51186-1G PC51186-250MG PC51186-5G
APOSPC51186-1GEA 131 EUR
APOSPC51186-1G APOSPC51186-250MG APOSPC51186-5G
5-Bromo-3-fluoro-2-hydroxybenzoic acid 97%
5-Bromo-3-fluoro-2-hydroxybenzoic acid
Formule: C₇H₄BrFO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06203615
CAS nummer: 251300-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR